Learn More
Biotium EV stains Cell and Organelle Stains Exosome EV stains ExoBrite 490 515 1/EA
Catalog No. 5000501103
Encompass_Preferred
ExoBrite Annexin EV Staining Kits are designed for staining EVs and exosomes for detection by flow cytometry This optimized formulation of Annexin V binds to phosphatidylserine in the EV membrane with bright signal and low background Ex/Em 490/516 nm 100 labelings
Catalog No. 50-005-01103 Supplier Biotium Supplier No. 30120T
ExoBrite Annexin EV Staining Kits are designed for staining EVs and exosomes for detection by flow cytometry This optimized formulation of Annexin V binds to phosphatidylserine in the EV membrane with bright signal and low background Ex/Em 490/516 nm 100 labelings