Learn More
Biotium EV stains Cell and Organelle Stains Exosome EV stains ExoBrite 555 575 1/EA
Catalog No. 5000501575
Encompass_Preferred
ExoBrite 555/575 True EV Membrane Stain is a membrane binding dye that stains EVs with full coverage and broad compatibility These dyes have improved solubility resulting in lower background and better signal to noise in EV detection compared to other membrane dyes 500X in DMSO 100 uL
Catalog No. 50-005-01575 Supplier Biotium Supplier No. 30130T
ExoBrite 555/575 True EV Membrane Stain is a membrane binding dye that stains EVs with full coverage and broad compatibility These dyes have improved solubility resulting in lower background and better signal to noise in EV detection compared to other membrane dyes 500X in DMSO 100 uL