Learn More
Biotium EV stains Cell and Organelle Stains Exosome EV stains ExoBrite 650 665 1/EA
Supplier:  Biotium 30122T
Encompass_Preferred
ExoBrite Annexin EV Staining Kits are designed for staining EVs and exosomes for detection by flow cytometry This optimized formulation of Annexin V binds to phosphatidylserine in the EV membrane with bright signal and low background Ex/Em 652/668 nm 100 labelings
Catalog No. 50-005-01332