Learn More
Biotium EV stains Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates ExoBrite 410 450 1/EA
Catalog No. 5000500085
Encompass_Preferred
ExoBrite WGA EV Staining Kits are designed for staining EVs and exosomes for detection by flow cytometry This optimized formulation of Wheat Germ Agglutinin (WGA) binds to carbohydrates in the EV membrane for bright staining Ex/Em 411/452 nm 500 labelings
Catalog No. 50-005-00085 Supplier Biotium Supplier No. 30123
ExoBrite WGA EV Staining Kits are designed for staining EVs and exosomes for detection by flow cytometry This optimized formulation of Wheat Germ Agglutinin (WGA) binds to carbohydrates in the EV membrane for bright staining Ex/Em 411/452 nm 500 labelings