Learn More
Biotium EV stains Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates ExoBrite 490 515 1/EA
Catalog No. 5000501333
Encompass_Preferred
ExoBrite WGA EV Staining Kits are designed for staining EVs and exosomes for detection by flow cytometry This optimized formulation of Wheat Germ Agglutinin (WGA) binds to carbohydrates in the EV membrane for bright staining Ex/Em 490/516 nm 500 labelings
Catalog No. 50-005-01333 Supplier Biotium Supplier No. 30124
ExoBrite WGA EV Staining Kits are designed for staining EVs and exosomes for detection by flow cytometry This optimized formulation of Wheat Germ Agglutinin (WGA) binds to carbohydrates in the EV membrane for bright staining Ex/Em 490/516 nm 500 labelings