Learn More
Biotium EV stains Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates ExoBrite 640 660 1/EA
Catalog No. 5000499733
Encompass_Preferred
ExoBrite WGA EV Staining Kits are designed for staining EVs and exosomes for detection by flow cytometry This optimized formulation of Wheat Germ Agglutinin (WGA) binds to carbohydrates in the EV membrane for bright staining Ex/Em 642/663 nm 500 labelings
Catalog No. 50-004-99733 Supplier Biotium Supplier No. 30126
ExoBrite WGA EV Staining Kits are designed for staining EVs and exosomes for detection by flow cytometry This optimized formulation of Wheat Germ Agglutinin (WGA) binds to carbohydrates in the EV membrane for bright staining Ex/Em 642/663 nm 500 labelings