Learn More
FUJIFILM Biosciences EV-Up MSC EV Production Sup AF
Catalog No. 5000450248
Encompass_Preferred
EV-Up MSC EV Production Supplemet AF is a supplement set used to prepare exosomes EVs production medium for mesenchymal stem cells MSC Mixture of this product and EV-Up MSC EV Production Supplement AF code No 298-84001 can use as the complete medium The products are serum-free and animal component-free available regardless of the type of growth medium The complete medium is available for MSCs derived from various tissue sources ex bone marrow umblical cord adipose tissue etc Enhances MSC-derived EV productionAnimal component-free AFSupplied as a two-part kit Supplement A and BSterility and mycoplasma tested For research use only RUO
Catalog No. 50-004-50248 Supplier FUJIFILM Biosciences Supplier No. 29884001
EV-Up MSC EV Production Supplemet AF is a supplement set used to prepare exosomes EVs production medium for mesenchymal stem cells MSC Mixture of this product and EV-Up MSC EV Production Supplement AF code No 298-84001 can use as the complete medium The products are serum-free and animal component-free available regardless of the type of growth medium The complete medium is available for MSCs derived from various tissue sources ex bone marrow umblical cord adipose tissue etc Enhances MSC-derived EV productionAnimal component-free AFSupplied as a two-part kit Supplement A and BSterility and mycoplasma tested For research use only RUO