Learn More
FUJIFILM Biosciences EV-UpEV Production Basal Mediu
Catalog No. 5000450334
Encompass_Preferred
EV-Up EV Production Basal Medium for MSC AF is a serum-free and animal component-free liquid medium for producing extracellular vesicles EVs with high bioactivity from MSCs while maintaining viability It is suitable for MSCs derived from various tissue sources
Catalog No. 50-004-50334 Supplier FUJIFILM Biosciences Supplier No. 05309451
EV-Up EV Production Basal Medium for MSC AF is a serum-free and animal component-free liquid medium for producing extracellular vesicles EVs with high bioactivity from MSCs while maintaining viability It is suitable for MSCs derived from various tissue sources