Learn More
Labtech Inc EV400VAC motor-lift rotary evaporator with built-in vacuum controller, Speed Range: 20 - 260 rmp, Temperature range: Ambient - 180℃, Temp. Control Mode: PID, Heating power: 1300W, Bath material: PTFE coating/SS, Power Supply: 115V, 60Hz
Catalog No. 5000496190
Encompass_Preferred
EV400VAC motor-lift rotary evaporator with built-in vacuum controller, Speed Range: 20 - 260 rmp, Temperature range: Ambient - 180℃, Temp. Control Mode: PID, Heating power: 1300W, Bath material: PTFE coating/SS, Power Supply: 115V, 60Hz
Catalog No. 50-004-96190 Supplier Labtech Inc Supplier No. LEV400VAC
EV400VAC motor-lift rotary evaporator with built-in vacuum controller, Speed Range: 20 - 260 rmp, Temperature range: Ambient - 180℃, Temp. Control Mode: PID, Heating power: 1300W, Bath material: PTFE coating/SS, Power Supply: 115V, 60Hz