Learn More
Sino Biological Human Enterovirus 71 VP0 Protein (His & GST Tag)
Supplier:  Sino Biological 40013V20B100
Encompass_Preferred
A DNA sequence encoding the amino acids (Met 1-Gln 323) of human enterovirus 71 genome polyprotein (Q66478-1), corresponding to the protein VP0 precusor, a component of immature procapsids, was fused with the N-terminal polyhistidine tag followed by a GST tag
Catalog No. 50-255-3766