Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ EVA Foam Ice Pans and Buckets with Lids

Catalog No. 03395151 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
1 L
2.5 L
4 L
9 L
Color:
Blue
Green
Purple
Includes:
Bucket with Lid
Pan with Lid
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Color Includes
03-395-151 1 L Green Pan with Lid
03-395-150 1 L Blue Pan with Lid
03-395-152 1 L Purple Pan with Lid
03-395-153 4 L Blue Pan with Lid
03-395-154 4 L Green Pan with Lid
03-395-155 4 L Purple Pan with Lid
03-395-156 9 L Blue Pan with Lid
03-395-157 9 L Green Pan with Lid
03-395-158 9 L Purple Pan with Lid
03-395-159 2.5 L Blue Bucket with Lid
03-395-160 2.5 L Green Bucket with Lid
03-395-161 2.5 L Purple Bucket with Lid
03-395-162 4 L Blue Bucket with Lid
03-395-163 4 L Green Bucket with Lid
03-395-164 4 L Purple Bucket with Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 03-395-151
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

removable lid.

Fisherbrand™EVA Foam Ice Pans and Buckets with Lids provide excellent insulation for any benchtop cryogenic application, without the mess!

  • Sweat-proof ethylene vinyl acetate will not leak
  • Compatible with ice, dry ice, liquid nitrogen, alcohol, and saline solutions
  • Bases and lids feature glove-friendly handles
  • Cleanup is quick and easy with molded pour spouts

Specifications

For Use With (Equipment) Ice, dry ice, liquid nitrogen, alcohol, and saline solutions
Includes Pan with Lid
Description Pan with Lid
Material EVA
Product Type Cryo-Safe Cold Box
Quantity 1
Color Green
Capacity (Metric) 1 L
Autoclavable Not Autoclavable
Temperature Control No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.