Learn More
Bullard™ EVA Pandemic PAPR System
SDP
Catalog No. 17986610 Shop All Bullard Company Products
Change view
Click to view available options
With 20TJDB single bib Tychem hood with acetate lens
Catalog No.
17-986-610 With 20TJDB single bib Tychem hood with acetate lens
1 options
Catalog No. 17-986-610 Supplier Bullard™ Supplier No. EVA20TJDB
Cutting-edge Powered Air-Purifying Respirator (PAPR) technology designed to provide constant air flow in a compact, streamlined form for added comfort
  • Lightweight design
  • Three different system configurations to meet most needs
  • NIOSH approved for use with DuPont™ Tychem™ CC20 series of hoods
Certifications
NIOSH
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.