Learn More
Bullard™ EVA PAPR System, Double Bib Hoods For Head Protection
SDP
Catalog No. 17989613 Shop All Bullard Company Products
Change view
Click to view available options
Facepiece/Headtop Model:
Tychem QC hood
Catalog No. Facepiece/Headtop Model
17-989-613 Tychem QC hood
1 options
Catalog No. 17-989-613 Supplier Bullard™ Supplier No. EVA20TICHT
Add to Cart
Add to Cart
For use with hard hats, EVA system includes PAPR, EVA blower, battery, charger, breathing tube. Choose from Tychem QC or Tychem SL double bib hoods.
  • Uniquely designed, streamlined form makes PAPR lightweight, eliminates bulky corners that can dig into the wearer's back, and balances the unit's weight for maximum comfort
  • Easy-to-read fuel gauge displays remaining battery capacity at a glance
  • Battery status can be checked at any time during use
  • Bullard Active Flow Technology automatically responds to a worker's need for more or less air flow
  • Two distinct audible alarms notify wearers of low battery and low air flow
  • Alarms are designed so the sound of the alarm travels through the breathing tube directly to the wearer's ear
  • Powered by a long-lasting 10,000-hour brushless motor
  • Single filter design for easy use
  • Filter locking tab notifies the user that the filter is in place with both a visual indication and audible click
Specifications
Battery Life 7 to 8 hours
Belt Material Foam
Product Type PAPR Hard Hat System
Facepiece/Headtop Model Tychem QC hood
Includes PAPR system, EVA blower, battery, charger, breathing tube
Material (Lens) Acetate
Model EVA20TICHT
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.