Learn More
Medchemexpress LLC HY-13327 5mg Medchemexpress, Evacetrapib CAS:1186486-62-3 Purity:>98%
SDP
Catalog No. 502081334
Encompass_Preferred
Medchemexpress, HY-13327 5mg Evacetrapib CAS:1186486-62-3 Evacetrapib is a potent and selective of CETP inhibitor, which inhibits human recombinant CETP protein (IC50 5.5 nM) and CETP activity in human plasma (IC50 36 nM) in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1334 Supplier Medchemexpress LLC Supplier No. HY133275MG
Medchemexpress, HY-13327 5mg Evacetrapib CAS:1186486-62-3 Evacetrapib is a potent and selective of CETP inhibitor, which inhibits human recombinant CETP protein (IC50 5.5 nM) and CETP activity in human plasma (IC50 36 nM) in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.