Learn More
Selleck Chemical LLC Evacetrapib (LY2484595) S2925-5mg
SDP
Catalog No. 503111695
Encompass_Preferred
Evacetrapib (LY2484595) is a potent and selective inhibitor of CETP with IC50 of 5 5 nM elevates HDL cholesterol without increases in aldosterone or blood pressure Phase 3
Catalog No. 50-311-1695 Supplier Selleck Chemical LLC Supplier No. S29255MG
Evacetrapib (LY2484595) is a potent and selective inhibitor of CETP with IC50 of 5 5 nM elevates HDL cholesterol without increases in aldosterone or blood pressure Phase 3