Learn More
Perkin Elmer US LLC Evacuable KBr Die O-Ring - Qty. 2
Catalog No. 502113501
Encompass_Preferred
The Evacuable Potassium Bromide Die prepares KBr pellets, 13 mm in diameter, of the highest quality and clarity. A pellet is formed inside an evacuated chamber and pressed between a highly polished anvil and plunger. This long-lasting die is precisely constructed to produce pellets of uniform quality.
Catalog No. 50-211-3501 Supplier Perkin Elmer US LLC Supplier No. 04800514
The Evacuable Potassium Bromide Die prepares KBr pellets, 13 mm in diameter, of the highest quality and clarity. A pellet is formed inside an evacuated chamber and pressed between a highly polished anvil and plunger. This long-lasting die is precisely constructed to produce pellets of uniform quality.