Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ready America Evacuation Kit
SDP

Catalog No. 17985140
Change view
Click to view available options
Weight (English):
5.3 lb.
7.8 lb.
10.6 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
17-985-140 7.8 lb.
17-985-139 5.3 lb.
17-985-141 10.6 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-985-140 Supplier Ready America Supplier No. 77150
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Three day emergency essentials for Pet with 5 year shelf-life

  • Emergency essentials you'll need for your pet in an emergency
  • 3 day emergency Kit
  • Evacuation Kit are also available for Cat and Dog
  • Emergency kit contains more than 10 essentials
  • 5 Year Shelf-life

Specifications

Product Type Pet Evacuation Kit
Dimensions (L x W x H) 16 x 10 x 10 in.
Weight (English) 7.8 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.