Learn More
Med Vet International Surgical Smoke Evacuator Starter Package Acu-Evac IE, STARTER-IE1, Each
Catalog No. 503045863
Encompass_Preferred
Acuderm's all purpose surgical smoke evacuator Suitable for use with CO2, erbium, hair removal and other laser or electrosurgical units in open procedures Patented 3 Inch clear hyperbolic horn with 1.3 Inch collection nozzle 98% Coefficient of entry 4% Loss, fraction of velocity head For maximum collection, hold within 3 Inch field of collection Front end filter loads within seconds, rotary variable air flow dial, variety of mounting options ULPA/Carbon filter, rated at 99.99997% at 0.01 micron and MPPS 0.12 micron main filter Sealed, closed system housing Electronic timer - 16 hours, plus low flow sensor/full filter alert AC Motor 63 Inch H2O static suction Non-corrosive/non-conductive cabinet 8 Foot Hospital grade cord/plug
Catalog No. 50-304-5863 Supplier Med Vet International Supplier No. 1010364EA
Add to Cart
Add to Cart
Request a Quote
Acuderm's all purpose surgical smoke evacuator Suitable for use with CO2, erbium, hair removal and other laser or electrosurgical units in open procedures Patented 3 Inch clear hyperbolic horn with 1.3 Inch collection nozzle 98% Coefficient of entry 4% Loss, fraction of velocity head For maximum collection, hold within 3 Inch field of collection Front end filter loads within seconds, rotary variable air flow dial, variety of mounting options ULPA/Carbon filter, rated at 99.99997% at 0.01 micron and MPPS 0.12 micron main filter Sealed, closed system housing Electronic timer - 16 hours, plus low flow sensor/full filter alert AC Motor 63 Inch H2O static suction Non-corrosive/non-conductive cabinet 8 Foot Hospital grade cord/plug