Learn More
Med Vet International Dynarex High Volume Evacuator Tip, White, 1000/Case
Catalog No. 502831889
Encompass_Preferred
Durable, smooth, and a standard for any dental practice. Designed to effectively suction saliva, water, and debris during dental procedures Features an adaptable double-ended tip for vented or non-vented use Crafted from premium-quality, durable materials for optimal reliability  Features smooth, rounded edges for greater patient comfort  Conveniently sized to fit standard HVE valves Not made with natural rubber latex
Catalog No. 50-283-1889 Supplier Med Vet International Supplier No. 2215
Durable, smooth, and a standard for any dental practice. Designed to effectively suction saliva, water, and debris during dental procedures Features an adaptable double-ended tip for vented or non-vented use Crafted from premium-quality, durable materials for optimal reliability  Features smooth, rounded edges for greater patient comfort  Conveniently sized to fit standard HVE valves Not made with natural rubber latex