Learn More
LAMDA BIOTECH INC EvaGreen Direct-qPCR Kit
Catalog No. 502045787
Encompass_Preferred
EvaGreen Direct-qPCR Kit contains all the reagents needed for a quick preparation of genomic DNA and the qPCR master mix for SYBR green real-time PCR assay. Any type of tissues can be used for this kit, such as mouse tails and nail tips, and other animal tissues and cells; plant leaf, root or seeds; bacteria, fungi and other samples. The fluorescent dye can be used in the same way as SYBR green but with a higher sensitivity and heat stability.
Catalog No. 50-204-5787 Supplier LAMDA BIOTECH INC Supplier No. D502R
EvaGreen Direct-qPCR Kit contains all the reagents needed for a quick preparation of genomic DNA and the qPCR master mix for SYBR green real-time PCR assay. Any type of tissues can be used for this kit, such as mouse tails and nail tips, and other animal tissues and cells; plant leaf, root or seeds; bacteria, fungi and other samples. The fluorescent dye can be used in the same way as SYBR green but with a higher sensitivity and heat stability.