Learn More
Biotium Fast EvaGreen Master Mix for qPCR(200 rxn)
Catalog No. NC0161507
Encompass_Preferred
Fast Evagreen qPCR Master Mix is a 2X qPCR master mix suitable for fast cycling protocols. The reagent system uses two of our breakthrough PCR technologies, Evagreen qPCR dye and Cheetah™ Taq hotstart DNA polymerase, to deliver superior results when compared to other commercial master mixes. It is supplied with a separate vial of ROX reference dye, for use with high ROX, low ROX, or no ROX instruments.For sensitive, affordable qPCR  reactions we now recommend our new and improved Forget-Me-Not™ EvaGreen qPCR Master Mixes.EvaGreen dye and applications are covered under granted and pending US and international patents. Cheetah™ Taq is covered by pending US and international patents.
Catalog No. NC0161507 Supplier Biotium Supplier No. 31003
Fast Evagreen qPCR Master Mix is a 2X qPCR master mix suitable for fast cycling protocols. The reagent system uses two of our breakthrough PCR technologies, Evagreen qPCR dye and Cheetah™ Taq hotstart DNA polymerase, to deliver superior results when compared to other commercial master mixes. It is supplied with a separate vial of ROX reference dye, for use with high ROX, low ROX, or no ROX instruments.For sensitive, affordable qPCR  reactions we now recommend our new and improved Forget-Me-Not™ EvaGreen qPCR Master Mixes.EvaGreen dye and applications are covered under granted and pending US and international patents. Cheetah™ Taq is covered by pending US and international patents.