Learn More
Bullard™ EVAHL PAPR Set with PAPRFC3 HEPA Filter
SDP
Catalog No. 17623664 Shop All Bullard Company Products
Change view
Click to view available options
:
With PAPRFC3 HEPA Filter
Catalog No.
17-623-664 With PAPRFC3 HEPA Filter
1 options
Catalog No. 17-623-664 Supplier Bullard™ Supplier No. EVAHLSTLN2KFC3
Only null left
Add to Cart
Add to Cart
Offers cutting edge hazardous location blower unit with the state-of-the-art Sightline hood. The STLN2K offers unparalleled comfort with 320 degree unobstructed vision and tremendous overall comfort.
Unique 320 degree vision to maximize vision and reduce claustrophobia; beveled bib to guarantee proper fit and simplify donning process; three channel air flow distribution and innovative drawstring / clip combination make the Sightline a great choice for all pharmaceutical operators.
  • PAFLEXBR breathing tube contours to all body types and eliminates any bowing effect. Blower unit provides up to 8.5 CFM airflow.
  • EVAHL blower features two speeds, dual alarms, and visual fuel gauge.
  • Comfortable belt is easy to clean. Unique flex breathing tube contours to the operator body and reduces SKUs.
Specifications
Color Black, White
Filter Type HEPA
Product Type PAPR Assembly with Hood
Size One Size