Learn More
Bullard™ EVAHL PAPR Set
SDP
Catalog No. 17623660 Shop All Bullard Company Products
Change view
Click to view available options
Bullard™ Blower and Sightline hood
Catalog No.
17-623-660 Bullard™ Blower and Sightline hood
1 options
Catalog No. 17-623-660 Supplier Bullard™ Supplier No. EVAHLSTLN2K
Add to Cart
Add to Cart
EVAHLSTLN2K offers cutting edge hazardous location blower unit with the state-of-the-art Sightline hood. The STLN2K offers unparalled comfort with 320 degree unobstructed vision and tremendous overall comfort. Unique flex breathing tube reduces SKUs and adds to experience.
Unique 320 degree vision to maximize vision and reduce claustrophobia; beveled bib to guarnateee proper fit and simplify donning process; three channel air flow distribution and innovative drawstring / clip combination make the Sightline a great choice for all pharmaceutical operators.
  • PAFLEXBR breating tube contours to all body types and eliminates any bowing effect. Blower unit provides up to 8.5 CFM airflow.
  • EVAHL blower features two speeds, dual alarms, and visual fuel gauge.
  • Comfortable belt is easy to clean. Unique flex breathing tube contours to the operator body and reduces SKUs.
Specifications
Color Black, White
Filter Type Available with a High Efficiency HEPA filter, Organic Vapor Cartridge/Filter Combo, or Acid Gas Cartridge/Filter Combo
Product Type PAPR Kit
Size One Size