Learn More
Sigma Aldrich Fine Chemicals Biosciences Evans Blue Dye content >=75 % | 314-13-6 | MFCD00004021 | 50G
Catalog No. NC0789432
Encompass
Evans Blue Dye content >=75 % | Mol Wt: 960.81 | 314-13-6 | MFCD00004021 | 50G
Catalog No. NC0789432 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E212950G
Evans Blue Dye content >=75 % | Mol Wt: 960.81 | 314-13-6 | MFCD00004021 | 50G