Learn More
Abcam Evans blue, L-glutamate uptake inhibitor, 1G
Catalog No. 502707183
Encompass_Preferred
MW 960.8 Da, Purity >95%. Potent inhibitor of L-glutamate uptake via the membrane bound excitatory amino acid transporter (EAAT). Able to antagonize AMPA and kainate receptor mediated currents (IC₅₀ values are 220 and 150 nM, respectively). Binds with high affinity to serum albumin.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7183 Supplier Abcam Supplier No. AB1208691G
MW 960.8 Da, Purity >95%. Potent inhibitor of L-glutamate uptake via the membrane bound excitatory amino acid transporter (EAAT). Able to antagonize AMPA and kainate receptor mediated currents (IC₅₀ values are 220 and 150 nM, respectively). Binds with high affinity to serum albumin.
The product is subject to the following: Abcam Restricted Use Statement