Learn More
Abcam Evans blue (mM/ml), L-glutamate uptake inhibitor, 100MM_ML
Catalog No. 502704506
Encompass_Preferred
MW 960.8 Da, Purity >95%. Potent inhibitor of L-glutamate uptake via the membrane bound excitatory amino acid transporter (EAAT). Able to antagonize AMPA and kainate receptor mediated currents (IC₅₀ values are 220 and 150 nM, respectively). Binds with high affinity to serum albumin.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4506 Supplier Abcam Supplier No. AB146727100MM_ML
MW 960.8 Da, Purity >95%. Potent inhibitor of L-glutamate uptake via the membrane bound excitatory amino acid transporter (EAAT). Able to antagonize AMPA and kainate receptor mediated currents (IC₅₀ values are 220 and 150 nM, respectively). Binds with high affinity to serum albumin.
The product is subject to the following: Abcam Restricted Use Statement