Learn More
Description
Evans blue is a non-permeating dye. In presence of plasma membrane damage, the dye enters in the cytoplasm and nucleus, thereby staining them blue. Evans blue is used for checking cell viability. It is also used to study BBB (blood-brain barrier) permeability. The dye binds to albumin and hence indicates BBB breakdown to protein.
Specifications
Specifications
| Chemical Name or Material | Evans Blue |
| CAS | 314-13-6 |
| Molecular Weight (g/mol) | 960.81 g/mol |
| Percent Purity | ≥75 % |
| Quantity | 50 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.