Learn More
Chemglass Life Sciences Dish, Evaporating, 50mL, PTFE, Flat Form
SDP
Catalog No. 501656791
Encompass_Preferred
Isostatically molded in pure, FDA approved, PTFE. Totally chemically resistant and inert. Super smooth internal finish.
Catalog No. 50-165-6791 Supplier Chemglass Life Sciences Supplier No. CG8276F02
Isostatically molded in pure, FDA approved, PTFE. Totally chemically resistant and inert. Super smooth internal finish.