Learn More
Technical Glass Products Inc Clear Fused Quartz Evaporating Dish 100ML Capacity
SDP
Catalog No. NC1255438
Encompass_Preferred
Clear Fused Quartz Evaporating Dish 100ML Capacity 80mm Diameter x 40mm Height
Catalog No. NC1255438 Supplier Technical Glass Products Inc Supplier No. ED100
Clear Fused Quartz Evaporating Dish 100ML Capacity 80mm Diameter x 40mm Height