Learn More
Technical Glass Products Inc Clear Fused Quartz Evaporating Dish 150ML Capacity
SDP
Catalog No. NC0198703
Encompass
Clear Fused Quartz Evaporating Dish 150ML Capacity 90mm Diameter x 45mm Height
Catalog No. NC0198703 Supplier Technical Glass Products Inc Supplier No. ED150
Clear Fused Quartz Evaporating Dish 150ML Capacity 90mm Diameter x 45mm Height