Learn More
Cole-Parmer Essentials Evaporating Dish, porcelain, round form, 250 mL, 6/pk
Catalog No. NC1828824 Shop All Cole-Parmer Products
Encompass_Preferred
Round-form evaporating dishes feature lip and spout. Capacities up to 250 mL (17816-10 through 17816-15) are glazed inside and out. Capacities 385 mL and greater (17816-16 through 17816-21) are glazed inside and partially glazed outside.
Catalog No. NC1828824 Supplier Cole-Parmer Supplier No. 1781615
Round-form evaporating dishes feature lip and spout. Capacities up to 250 mL (17816-10 through 17816-15) are glazed inside and out. Capacities 385 mL and greater (17816-16 through 17816-21) are glazed inside and partially glazed outside.