Learn More
Chemglass Life Sciences 10L Flask, Evaporating, Large Scale, 100mm Flange, 335mm OAH
SDP
Catalog No. CG133702
Encompass_Preferred
10L Flask, Evaporating, Large Scale, 100mm Flange, 335mm OAH. Flasks are compatible with Buchi large scale rotary evaporators. Flasks to fit large scale rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation.
Catalog No. CG133702 Supplier Chemglass Life Sciences Supplier No. CG133702
10L Flask, Evaporating, Large Scale, 100mm Flange, 335mm OAH. Flasks are compatible with Buchi large scale rotary evaporators. Flasks to fit large scale rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation.