Learn More
Chemglass Life Sciences 20L Flask, Evaporating, Large Scale, 100mm Flange, 375mm OAH
SDP
Catalog No. CG133705
Encompass_Preferred
20L Flask, Evaporating, Large Scale, 100mm Flange, 375mm OAH. Flasks are compatible with Buchi large scale rotary evaporators. Flasks to fit large scale rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation.
Catalog No. CG133705 Supplier Chemglass Life Sciences Supplier No. CG133705
20L Flask, Evaporating, Large Scale, 100mm Flange, 375mm OAH. Flasks are compatible with Buchi large scale rotary evaporators. Flasks to fit large scale rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation.