Learn More
Heidolph™ Evaporating Flasks
SDP
Catalog No. 13878573 Shop All Heidolph North America Products
Change view
Click to view available options
Volume (Metric):
1000 mL
Catalog No. Volume (Metric)
13-878-573 1000 mL
1 options
Catalog No. 13-878-573 Supplier Heidolph™ Supplier No. 036301160
Add to Cart
Add to Cart
Heidolph™ Evaporating Flasks for use with Rotary Evaporator
  • Evaporating flasks come with a standard NS 24/40 joint size
  • Also available in joint size NS 29/32
Specifications
Description Round Bottom
Material Glass
For Use With (Equipment) Rotary Evaporator
Size NS 24/40
Taper Joint: NS 24/40
Volume (Metric) 1000 mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov