Learn More
Glas Col LLC Evaporator
SDP
Catalog No. NC1826663
Encompass_Preferred
Evaporator for 24 - 15mm vials, insert and gas inlet heated to 100 degrees C. 240V CE
Catalog No. NC1826663 Supplier Glas Col LLC Supplier No. 109A 1180245CE
Evaporator for 24 - 15mm vials, insert and gas inlet heated to 100 degrees C. 240V CE