Learn More
Nanoentek Inc EVE HT FL COUNTING PLATE
SDP
Catalog No. NC2581586
Encompass
  • Designed for high-throughput cell counting and viability analysis.
  • Facilitates processing of up to 48 samples per counting plate.
  • Optimizes workflow for cell line development and large-scale production.
  • Includes 20 counting plates per kit for efficient analysis.
  • Each counting plate features 48 channels.
  • Comes with 10 mixing well plates, each containing 96 wells.
  • Includes 20 reservoirs for sample management.
  • Requires a small sample volume of 20 µL per channel.
Catalog No. NC2581586 Supplier Nanoentek Inc Supplier No. EVFL020
  • Designed for high-throughput cell counting and viability analysis.
  • Facilitates processing of up to 48 samples per counting plate.
  • Optimizes workflow for cell line development and large-scale production.
  • Includes 20 counting plates per kit for efficient analysis.
  • Each counting plate features 48 channels.
  • Comes with 10 mixing well plates, each containing 96 wells.
  • Includes 20 reservoirs for sample management.
  • Requires a small sample volume of 20 µL per channel.