Learn More
Medchemexpress LLC HY-17612 5mg Medchemexpress, Evenamide CAS:1092977-61-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY176125MG
Encompass_Preferred
Medchemexpress, HY-17612 5mg Evenamide CAS:1092977-61-1 Evenamide (NW-3509), a sodium channel blocker, shows efficacy in a broad spectrum of rodent models of psychosis, mania, depression, and aggressiveness. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9999