Learn More
TA Instruments Event Cable FTIR, Simultaneous Start TGA or SDT Module and FTIR, Compatible with Thermo Nicolet 6700 iS10 FTIR, Included in TGA FTIR Interface Kit
Catalog No. 5000580606 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Event Cable for FTIR allows simultaneous start of a TGA or SDT module and an FTIR spectrometer to initiate a combined TGA/FTIR experiment. It is compatible with Thermo Nicolet 6700 and iS10 FTIR models. The cable is also supplied in the TGA/FTIR Interface Kit.
Catalog No. 50-005-80606 Supplier TA Instruments Supplier No. 957481.901
Waters TA Instruments Event Cable for FTIR allows simultaneous start of a TGA or SDT module and an FTIR spectrometer to initiate a combined TGA/FTIR experiment. It is compatible with Thermo Nicolet 6700 and iS10 FTIR models. The cable is also supplied in the TGA/FTIR Interface Kit.