Learn More
Waters Corp Waters Cable Assembly, Event In/Out, HPLC
Catalog No. 50793497 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Event In/Out Cable Assembly is designed to connect external devices, such as manual injectors, to the 10-pin terminal strip located on the rear panel of the instrument.
  • Functions as an input/output cable
  • Designed for HPLC systems
  • Replaced as needed to maintain system performance
Catalog No. 50-793-497 Supplier Waters Corp Supplier No. WAT020321
The Event In/Out Cable Assembly is designed to connect external devices, such as manual injectors, to the 10-pin terminal strip located on the rear panel of the instrument.
  • Functions as an input/output cable
  • Designed for HPLC systems
  • Replaced as needed to maintain system performance