Learn More
Pyramex™ Ever-Lite™ Safety Eyewear
Catalog No. 19155844 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Anti-fog, Anti-scratch
Uncoated
Lens Tint:
Amber
Blue
Blue, Mirror
Clear
Dark Gray
Gray
Indoor/Outdoor, Mirror
Photochromic
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint
19-155-844 Anti-fog, Anti-scratch Clear
19-155-842 Uncoated Clear
19-155-843 Anti-fog Clear
19-155-845 Uncoated Gray
19-155-846 Anti-fog Gray
19-155-847 Anti-fog, Anti-scratch Gray
19-155-848 Uncoated Dark Gray
19-155-849 Anti-fog Dark Gray
19-155-850 Uncoated Photochromic
19-155-851 Uncoated Amber
19-155-852 Uncoated Blue
19-155-853 Anti-fog Blue
19-155-854 Uncoated Blue, Mirror
19-155-855 Uncoated Indoor/Outdoor, Mirror
19-155-856 Anti-fog Indoor/Outdoor, Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 19-155-844 Supplier Pyramex™ Supplier No. SB8610DTM
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Lightweight spectacle (22 grams) with a soft nosepiece for snug fit, semi-frame, and slim co-injected temples
  • Offers a clear panoramic view without obstruction
  • 8 base wraparound lens provides full front side protection
  • Scratch-resistant, polycarbonate lens provide 99.9% of UV protection
  • H2MAX creates a high performing, superior anti-fog coating that is durable enough to withstand repeated cleaning without losing adhesion
  • H2MAX ensures maximum clarity in the most extreme temperature and humidity conditions, and it also adds a layer to the lens that is abrasion, chemical, and UV-resistant
  • Specifications
    Lens Coating Anti-fog, Anti-scratch
    Lens Type H2MAX
    Certifications/Compliance ANSI Z87.1,CSA Z94.3-07
    Certifications/Compliance (Europe) EN-166
    Lens Tint Clear
    Product Type Safety Eyewear
    Frame Color Black
    Weight (Metric) 22 g
    Material Polycarbonate, TPR
    Disposable Reusable
    Lens Material Polycarbonate
    Frame Material Polycarbonate
    UV Protection U6
    Frame Type Half Frame