Learn More
Pyramex™ Ever-Lite™ Safety Eyewear
Catalog No. 19155856 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Amber
Blue
Blue, Mirror
Clear
Dark Gray
Gray
Indoor/Outdoor, Mirror
Photochromic
Lens Coating:
Anti-fog
Anti-fog, Anti-scratch
Uncoated
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating
19-155-856 Indoor/Outdoor, Mirror -
19-155-842 Clear -
19-155-843 Clear -
19-155-844 Clear Anti-fog, Anti-scratch
19-155-845 Gray -
19-155-846 Gray -
19-155-847 Gray Anti-fog, Anti-scratch
19-155-848 Dark Gray -
19-155-849 Dark Gray -
19-155-850 Photochromic -
19-155-851 Amber -
19-155-852 Blue Uncoated
19-155-853 Blue Anti-fog
19-155-854 Blue, Mirror -
19-155-855 Indoor/Outdoor, Mirror -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 19-155-856 Supplier Pyramex™ Supplier No. SB8680DT
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight spectacle (22 grams) with a soft nosepiece for snug fit, semi-frame, and slim co-injected temples
  • Offers a clear panoramic view without obstruction
  • 8 base wraparound lens provides full front side protection
  • Scratch-resistant, polycarbonate lens provide 99.9% of UV protection
  • H2MAX creates a high performing, superior anti-fog coating that is durable enough to withstand repeated cleaning without losing adhesion
  • H2MAX ensures maximum clarity in the most extreme temperature and humidity conditions, and it also adds a layer to the lens that is abrasion, chemical, and UV-resistant
  • Specifications
    Lens Type H2X
    Certifications/Compliance ANSI Z87.1
    Lens Tint Indoor/Outdoor, Mirror
    Frame Color Black
    Weight (Metric) 22 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate