Learn More
Biotium EverBrite Mounting Medium, w/o DAPI, 10 mL
Catalog No. NC1370565
Encompass_Preferred
A revolutionary antifade mounting medium optimally formulated for preserving fluorescence of our CF dyes and other fluorochromes.
Catalog No. NC1370565 Supplier Biotium Supplier No. 23001
A revolutionary antifade mounting medium optimally formulated for preserving fluorescence of our CF dyes and other fluorochromes.