Learn More
Barno Electronics Button Cell Type 357 Battery by Energizer, Pack of 1
SDP
Catalog No. NC0326253
Encompass_Preferred
1 X Button Cell Type 357 Battery by Energizer 4.5 out of 5 stars 8 customer reviews Type: Silver Oxide Battery is 1.55 V Has a capacity of 150 mah. Energizer multi-drain Drain. *Image provided by supplier may depict different quantity.
Catalog No. NC0326253 Supplier Barno Electronics Supplier No. 357
1 X Button Cell Type 357 Battery by Energizer 4.5 out of 5 stars 8 customer reviews Type: Silver Oxide Battery is 1.55 V Has a capacity of 150 mah. Energizer multi-drain Drain. *Image provided by supplier may depict different quantity.