Learn More
Enterprise Technology Solutions Eveready Battery Co Inc Flashlight,Intrinsically Safe,Waterproof,LED,OE
SDP
Supplier:  Enterprise Technology Solutions EVEENISHH25E
Encompass_Preferred
Go to work prepared with this Intrinsically Safe Handheld LED Flashlight. High-performance LED light is specifically designed to meet the needs of industrial trades. It is approved by global safety agencies to safely operate in environments with water, gas or dust. Waterproof flashlight keeps shining even after fully immersing in 1-meter deep water for 30 minutes. Rugged design is also great as an emergency light or camping flashlight. Beam reaches over 570' with an exceptional runtime of 65 hours. Battery-powered, impact-resistant light features a shatterproof lens and survives drop after drop up to 3'. Orange safety color is easy to spot on the job. Ribbed casing provides a secure grip. Built-in loop lets you secure it to a rig.
Catalog No. 50-008-31269