Learn More
Enterprise Technology Solutions Eveready Battery Co Inc LED Flashlight Combo, 3-47/50"x2-19/25"x6-1/5",4/PK AST
SDP
Supplier:  Enterprise Technology Solutions EVEEVM5511S
Encompass_Preferred
LED flashlight combo pack with super heavy-duty batteries features four flashlights in two sizes to provide 25 lumens of bright white light for a variety of tasks around your home or office. Ribbed casing provides both strength and a nonslip grip for reliable use in most conditions. Easy-to-operate push-button switch allows convenient, single-handed operation to keep your other hand free. Long-lasting LED bulbs provide up to 30 hours of runtime with AA batteries and 60 hours of runtime with D batteries.
Catalog No. 50-008-30687