Learn More
Honeywell North™ Everest A119R Hard Hat with Irvine Ranch Water District Logo
Catalog No. 19085595
Change view
Click to view available options
Full Brim
Catalog No.
19-085-595 Full Brim
1 options
Catalog No. 19-085-595 Supplier Honeywell North™ Supplier No. A119R03B1492
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Ideal for workers who are exposed to the risk of a blow to the side, front, or rear of the head. Also protects workers from the hazards associated with falling debris, rain, or the sun's UV rays.
TRUSTED_SUSTAINABILITY
Specifications
Width (English) 11.5 in.
Width (Metric) 29.21 cm
Certifications/Compliance Meets ANSI Z89.1-2009 and CSA Z94.1-2005 Type 1 Class E. A119R: Meets ANSI Z89.1-2009 and CSA Z94.1 Type 2 Class E
Color Orange
Includes Irvine Ranch Water District Logo
Material HDPE Shell
Model Everest A119R
Product Type Hard Hat
Size One Size Fits Most
Style Ratchet Adjustment
Suspension Type 6 Point
Ventilated Not Ventilated
Head Style Full Brim