Learn More
Caplugs™ Evergreen Polypropylene Column
Catalog No. 057239E Shop All Caplugs Products
Change view
Click to view available options
Length (Metric):
127 mm
Catalog No. Length (Metric)
05-723-9E 127 mm
1 options
Catalog No. 05-723-9E Supplier Caplugs™ Supplier No. 2083384060
Specifications
Type Chromatography Column
Endcapped Yes
For Use With (Application) Organic solvents and stronger Acids (except oxidizing Acids) and Alkalis
Length (Metric) 127 mm
Material Polypropylene
Includes Reservoir, end caps and Polyethylene filter matrix