Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caplugs™ Evergreen Scientific CryoSure™ Brand Cryogenic Vials without Caps

Catalog No. 055544 Shop All Caplugs Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
055544 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 055544 Supplier Caplugs™ Supplier No. 2203902080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

External thread, freestanding vials provide safe, secure, reliable storage for specimens in liquid nitrogen vapor-phase™

Recommended for storing cell cultures, blood/serum specimens, sperm, and other biological specimens at vapor phase™ liquid nitrogen temperatures (-320°F/-195°C).

  • External thread design is less susceptible to aerosol contamination than internally threaded vials, and have smoother engagement than ordinary 60° threads.
  • Ultra-clear polypropylene material is nearly transparent and provides superior tensile strength and resistance to gamma radiation sterilization.
  • Permanent marking areas are molded in for labeling and specimen identification, with volume graduations distinctly molded every 0.5mL.
  • Straight vertical inside walls with no shoulder result in a large accessible opening which makes processing and retrieval of samples convenient.

For use with Evergreen Scientific CryoSure Vial Caps (055584), sold separately.

Specifications

Color Clear
Material Polypropylene
Sterility Non-sterile
Volume (Metric) 1.5 mL
For Use With (Application) Vapor phase liquid nitrogen only
Thread Style External
Closure Type Not Included
Quantity 1000/Cs.

™Please use CryoSure vials in the vapor-phase only. Evergreen does not recommend the use of screwcap vials, including CryoSure vials, for immersion in liquid nitrogen or other liquified gases. Immersion in liquid nitrogen may cause leakage in CryoSure vials and also presents the potential danger of rapid pressure buildup and/or explosion when returned to room temperature.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.