Learn More
Caplugs™ Evergreen Scientific CryoSure™ Vials Caps
Catalog No. 055584 Shop All Caplugs Products
Change view
Click to view available options
Quantity:
200/Cs.
Catalog No. Quantity
05-558-4 200/Cs.
1 options
Catalog No. 05-558-4 Supplier Caplugs™ Supplier No. 3003900Y20
Add to Cart
Add to Cart
Leakproof closure without the use of a silicone gasket
Polyethlyene screwcaps feature a molded-in sealing ring for a rim-seal that offers sealing efficacy in excess of 99 percent.
  • Fit all CryoSure vials
  • Color: yellow
  • Nonsterile
Specifications
Closure Color Yellow
Material Polyethylene
Sterility Non-sterile
Closure Material Polyethylene
Closure Type Screw Cap
Quantity 200/Cs.