Learn More
Caplugs™ (Evergreen Scientific) Empty Histoware™ Vials
Catalog No. 055532 Shop All Caplugs Products
Change view
Click to view available options
Capacity (Metric):
60 mL
90 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
05-553-2 60 mL
055533 90 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-553-2 Supplier Caplugs™ (Evergreen Scientific) Supplier No. 2405601G80
Add to Cart
Add to Cart
Unfilled shatterproof vials for histology work
  • Formulated polypropylene provides unmatched vial clarity and tensile strength
  • Shatterproof if accidentally dropped
  • Green polyethylene screwcaps have molded-in sealing rings for virtually leakproof seals
Specifications
Capacity (Metric) 60 mL
Closure Color Green
Closure Material Polyethylene
Closure Type Screw Cap